Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Dichloroindole 98.0+%, TCI America™
SDP

Catalog No. p-7128552
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 121859-57-2
Molecular Formula C8H5Cl2N
Molecular Weight (g/mol) 186.04
MDL Number MFCD01075036
InChI Key ILINOHVVKWYAFM-UHFFFAOYSA-N
PubChem CID 10487776
IUPAC Name 5,6-dichloro-1H-indole
SMILES ClC1=C(Cl)C=C2C=CNC2=C1

Specifications

Melting Point 152°C
Quantity 1 g
Formula Weight 186.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dichloroindole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.