Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Dihydro-1,4-dithiin-2,3-dicarboximide 97.0+%, TCI America™
SDP

Catalog No. p-7127378
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 24519-85-5
Molecular Formula C6H5NO2S2
Molecular Weight (g/mol) 187.23
MDL Number MFCD01632185
InChI Key XNIVQWHLYNEVBN-UHFFFAOYSA-N
Synonym 2,3-Dihydro-1,4-dithiino[2,3-c]pyrrole-5,7-dione
PubChem CID 32477
IUPAC Name 2H,3H,5H,6H,7H-[1,4]dithiino[2,3-c]pyrrole-5,7-dione
SMILES O=C1NC(=O)C2=C1SCCS2

Specifications

Melting Point 217°C
Quantity 1 g
Formula Weight 187.23
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dihydro-1,4-dithiin-2,3-dicarboximide
RTECSNumber JO6955000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.