Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Dihydrouracil, 97%

Catalog No. p-7043654
Click to view available options
Quantity:
1 g
5 g

CAS: 504-07-4 | C4H6N2O2 | 114.10 g/mol

5,6-Dihydrouracil acts as an intermediate in the catabolism of uracil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,6-Dihydrouracil acts as an intermediate in the catabolism of uracil.

Solubility
Soluble in Sodium Hydroxide.

Notes
Incompatible with oxidizing agents.Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-07-4
Molecular Formula C4H6N2O2
Molecular Weight (g/mol) 114.10
MDL Number MFCD00006029
InChI Key OIVLITBTBDPEFK-UHFFFAOYSA-N
Synonym dihydrouracil, 5,6-dihydrouracil, hydrouracil, 5,6-dihydro-2,4-dihydroxypyrimidine, dihydropyrimidine-2,4 1h,3h-dione, dihydrouracile, 2,4 1h,3h-pyrimidinedione, dihydro, di-h-uracil, dihydro-2,4 1h,3h-pyrimidinedione, hydrouracil 8ci
PubChem CID 649
ChEBI CHEBI:15901
IUPAC Name 1,3-diazinane-2,4-dione
SMILES O=C1CCNC(=O)N1

Specifications

Melting Point 278°C to 280°C
Quantity 1 g
Beilstein 112496
Solubility Information Soluble in Sodium Hydroxide.
Formula Weight 114.1
Percent Purity 97%
Chemical Name or Material 5,6-Dihydrouracil
EINECSNumber 207-982-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.