Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Dimethyluracil 99.0+%, TCI America™
SDP

Catalog No. D11361G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11361G 1 g
D11365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D11361G Supplier TCI America Supplier No. D11361G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26305-13-5
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD00010708
InChI Key PZVLJGKJIMBYNP-UHFFFAOYSA-N
Synonym 2,4-Dihydroxy-5,6-dimethylpyrimidine, 5,6-Dimethyl-2,4-pyrimidinediol
PubChem CID 96031
IUPAC Name 5,6-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES CC1=C(C)C(=O)NC(=O)N1

Specifications

Color White
Quantity 1 g
Formula Weight 140.14
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dimethyluracil
RTECSNumber YQ9750000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.