Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4'-disulfonic acid disodium salt hydrate, 97+%

Catalog No. aal1060703
Click to view available options
Quantity:
1 g
5 g

CAS: 1264198-47-1 | C20H12N4Na2O6S2 | 514.44 g/mol

Reagent for determination of serum iron. Reagent for the simultaneous spectrophotometric determination of iron and copper. PDT is the non-sulfonated precursor to Ferrozine (FZ), a commonly used reagent for Fe(II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for determination of serum iron. Reagent for the simultaneous spectrophotometric determination of iron and copper. PDT is the non-sulfonated precursor to Ferrozine (FZ), a commonly used reagent for Fe(II).

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1264198-47-1
Molecular Formula C20H12N4Na2O6S2
Molecular Weight (g/mol) 514.44
MDL Number MFCD00717621
InChI Key DDIHLFCSEHAOJZ-UHFFFAOYSA-L
Synonym ferrozin, ferrozine, ferrozine disodium salt, unii-29j2ciw9pk, 29j2ciw9pk, 3-2-pyridyl-5,6-di 4-phenylsulfonic acid-1,2,4-triazine, disodium salt, 4,4'-3-2-pyridyl-triazine-5,6-diyl dibenzenesulfonic acid disodium salt, 4,4'-3-2-pyridinyl-1,2,4-triazine-5,6-diyl bis-benzenesulfonic acid, disodium salt, 4,4'-3-2-pyridinyl-1,2,4-triazine-5,6-diyl bis-benzenesulfonic acid,disodium salt, benzenesulfonic acid, 4,4'-3-2-pyridinyl-1,2,4-triazine-5,6-diyl bis-, disodium salt
PubChem CID 34127
IUPAC Name disodium 4-[3-(pyridin-2-yl)-5-(4-sulfonatophenyl)-1,2,4-triazin-6-yl]benzene-1-sulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=NN=C(N=C1C1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=CC=N1

Specifications

Quantity 1 g
Beilstein 5222087
Solubility Information Soluble in water.
Formula Weight 514.46 (Anhydrous)
Percent Purity 98%
Chemical Name or Material 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4'-disulfonic acid disodium salt hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 248-797-6
RTECSNumber DB7345000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.