Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,7-Difluoro-1-indanone, 97%

Catalog No. AAH5400206
Click to view available options
Quantity:
1 g
5 g

CAS: 84315-25-3 | C9H6F2O | 168.14 g/mol

5,7-Difluoro-1-indanone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,7-Difluoro-1-indanone is used as pharmaceutical intermediate.

Solubility
Sparingly soluble in water 0.081 g/L 25°C.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84315-25-3
Molecular Formula C9H6F2O
Molecular Weight (g/mol) 168.14
MDL Number MFCD09908161
InChI Key XWZGNWCKXLECBO-UHFFFAOYSA-N
Synonym 5,7-difluoro-1-indanone, 5,7-difluoro-2,3-dihydro-1h-inden-1-one, 1h-inden-1-one, 5,7-difluoro-2,3-dihydro, 5,7-difluoroindan-1-one, 1h-inden-1-one,5,7-difluoro-2,3-dihydro, acmc-20a83k, 5,7-difluoro-2,3-dihydro-1h-indene-1-one
PubChem CID 11789595
IUPAC Name 5,7-difluoro-2,3-dihydroinden-1-one
SMILES FC1=CC(F)=C2C(=O)CCC2=C1

Specifications

Melting Point 81°C
Quantity 5 g
Solubility Information Sparingly soluble in water 0.081g/L 25°C.
Formula Weight 168.14
Percent Purity 97%
Chemical Name or Material 5,7-Difluoro-1-indanone
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.