Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,7-Dihydroxy-4'-methoxyisoflavone, 98%

Catalog No. AA4385306
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 491-80-5 | C16H12O5 | 284.27 g/mol

It has putative benefits in dietary cancer prophylaxis. It has also been found to inhibit fatty acid amide hydrolase and to act as agonist of PPARgamma, nuclear receptor that is current pharmacological target for the treatment of diabetes type 2. It acts as an antineoplastic agent. It is a selective agonist at ER-β estrogen receptors, and may have chemopreventive efficacy against breast cancer. In line with its low activity at ER-α estrogen receptors, it is essentially devoid of uterotrophic activity. Biochanin A is also a ligand for the aryl hydrocarbon receptor (AhR). It reduces arterial resistance and enhances microcirculation perhaps via effects on potassium and/or calcium ion channels. Induction of sulfotransferases for xenobiotic detoxification has been proposed as a mechanism of its cancer preventive effects. It is a nitric oxide synthase inhibitor and apoptosis inducer

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 491-80-5
Molecular Formula C16H12O5
Molecular Weight (g/mol) 284.27
MDL Number MFCD00006839
InChI Key WUADCCWRTIWANL-UHFFFAOYSA-N
Synonym biochanin a, biochanin, 4'-methylgenistein, 5,7-dihydroxy-4'-methoxyisoflavone, 5,7-dihydroxy-3-4-methoxyphenyl-4h-chromen-4-one, genistein 4-methyl ether, pratensol, biochanine a, 5,7-dihydrox-4'-methoxyisoflavone, 4h-1-benzopyran-4-one, 5,7-dihydroxy-3-4-methoxyphenyl
PubChem CID 5280373
ChEBI CHEBI:17574
IUPAC Name 5,7-dihydroxy-3-(4-methoxyphenyl)chromen-4-one
SMILES COC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O

Specifications

Melting Point 210°C to 213°C
Quantity 5 g
Beilstein 278107
Solubility Information Soluble in water (<1mg/ml at 25°C),chloroform,methanol,DMSO (57mg/ml at 25°C),and ethanol (9mg/ml at 25°C).
Formula Weight 284.27
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5,7-Dihydroxy-4'-methoxyisoflavone
EINECSNumber 207-744-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.