Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,8-Dibromo-2,3-diphenylquinoxaline 98.0+%, TCI America™
SDP

Catalog No. D4461200MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg

Chemical Identifiers

CAS 94544-77-1
Molecular Formula C20H12Br2N2
Molecular Weight (g/mol) 440.138
InChI Key LELMSURPXIAXOW-UHFFFAOYSA-N
PubChem CID 22475901
IUPAC Name 5,8-dibromo-2,3-diphenylquinoxaline
SMILES C1=CC=C(C=C1)C2=NC3=C(C=CC(=C3N=C2C4=CC=CC=C4)Br)Br

Specifications

Melting Point 222°C
Color Yellow
Quantity 200 mg
Formula Weight 440.14
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5,8-Dibromo-2,3-diphenylquinoxaline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.