Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Acetamido-2-(pentafluoroethyl)pyridine, 96%, Thermo Scientific™

Catalog No. aah66076mc
Click to view available options
Quantity:
100 mg

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1580464-60-3
Molecular Formula C9H7F5N2O
Molecular Weight (g/mol) 254.16
InChI Key FTIYUAIKVLBBTH-UHFFFAOYSA-N
Synonym N-(6-Perfluoroethyl-3-pyridyl)acetamide
PubChem CID 99728620
IUPAC Name N-[6-(1,1,2,2,2-pentafluoroethyl)pyridin-3-yl]acetamide
SMILES CC(=O)NC1=CN=C(C=C1)C(C(F)(F)F)(F)F

Specifications

Quantity 100 mg
Formula Weight 254.16
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 5-Acetamido-2-(pentafluoroethyl)pyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.