Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Acetonyl-2-methoxybenzenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. A294825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A294825G 25 g
A29485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A294825G Supplier TCI America Supplier No. A294825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 116091-63-5
Molecular Formula C10H13NO4S
Molecular Weight (g/mol) 243.28
MDL Number MFCD07782136
InChI Key MQQJFLHZXQRKKJ-UHFFFAOYSA-N
Synonym 2-Methoxy-5-(2-oxopropyl)benzenesulfonamide
PubChem CID 14179759
IUPAC Name 2-methoxy-5-(2-oxopropyl)benzene-1-sulfonamide
SMILES COC1=C(C=C(CC(C)=O)C=C1)S(N)(=O)=O

Specifications

Melting Point 196°C
Quantity 25 g
Formula Weight 243.28
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Acetonyl-2-methoxybenzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.