Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Acetyl-2-methoxypyridine 98.0+%, TCI America™
SDP

Catalog No. A27975G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 213193-32-9
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD05664350
InChI Key RYOQZXOVBJIUSX-UHFFFAOYSA-N
Synonym 5-acetyl-2-methoxypyridine, 1-6-methoxypyridin-3-yl ethanone, 1-6-methoxypyridin-3-yl ethan-1-one, 1-6-methoxy-3-pyridyl ethanone, ethanone, 1-6-methoxy-3-pyridinyl, 3-acetyl-6-methoxypyridine, 1-6-methoxy-3-pyridinyl ethanone, pubchem16770, acmc-209fj4, 2-methoxy-5-acetylpyridine
PubChem CID 10534837
IUPAC Name 1-(6-methoxypyridin-3-yl)ethanone
SMILES CC(=O)C1=CN=C(C=C1)OC

Specifications

Melting Point 53°C
Color Yellow
Quantity 5 g
Formula Weight 151.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Acetyl-2-methoxypyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.