Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Acetyl-2-thiopheneboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193887 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Specifications

Chemical Name or Material 5-Acetyl-2-thiopheneboronic Acid (contains varying amounts of Anhydride)
CAS 206551-43-1
Molecular Formula C6H7BO3S
Quantity 5 g
MDL Number MFCD01075681
Synonym 5-acetylthiophene-2-boronic acid, 5-acetyl-2-thiopheneboronic acid, 5-acetyl-2-thienylboronic acid, 5-acetylthiophen-2-yl boronic acid, 5-acetyl-2-thienyl boronic acid, 5-acetylthiophen-2-boronic acid, boronic acid, 5-acetyl-2-thienyl, 2-acetylthien-5-yl boronic acid, 5-acetylthien-2-yl boronic acid, 1-5-dihydroxyboranyl thiophen-2-yl ethan-1-one
InChI Key DCNMATSPQKWETQ-UHFFFAOYSA-N
SMILES B(C1=CC=C(S1)C(=O)C)(O)O
IUPAC Name (5-acetylthiophen-2-yl)boronic acid
Molecular Weight (g/mol) 169.989
PubChem CID 4143377
Formula Weight 169.99
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.