Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Acetylsalicylic acid, 98+%

Catalog No. AAA1166306
Click to view available options
Quantity:
5 g
25 g
100 g

Catalog No. AAA1166306

Supplier: Thermo Scientific Chemicals A1166306

Only null left
Add to Cart

CAS: 13110-96-8 | C9H8O4 | 180.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13110-96-8
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00013978
InChI Key NZRDKNBIPVLNHA-UHFFFAOYSA-N
PubChem CID 83151
IUPAC Name 5-acetyl-2-hydroxybenzoic acid
SMILES CC(=O)C1=CC(=C(C=C1)O)C(=O)O

Specifications

Melting Point 214°C to 216°C
Flash Point 250°C (482°F)
Quantity 5 g
Beilstein 2096968
Sensitivity Moisture sensitive
Synonym 5-acetylsalicylic acid, 3-acetyl-6-hydroxybenzoic acid, 5-acetyl-2-hydroxy-benzoic acid, benzoic acid, 5-acetyl-2-hydroxy, 5-acetyl salicylic acid, acmc-1boxk, 5-acetylsalicylicacid, 5-acetyl-2-hydroxybenzoic acid, benzoic acid,5-acetyl-2-hydroxy
Formula Weight 180.15
Percent Purity ≥98%
Chemical Name or Material 5-Acetylsalicylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-037-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.