Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Acetylvaleric acid, 90+%, technical

Catalog No. ac449530050
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 3128-07-2 | C7H10O3 | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3128-07-2
Molecular Formula C7H10O3
Molecular Weight (g/mol) 144.17
InChI Key IZOQMUVIDMLRDC-UHFFFAOYSA-N
Synonym 5-acetylvaleric acid, heptanoic acid, 6-oxo, 5-aceto valeric acid, 6-oxo-heptanoic acid, unii-4u902i8qzo, 5-acetylvalericacid, 6-oxoenanthic acid, 6-ketoheptanoic acid, 5-acetylpentanoic acid, #
PubChem CID 98810
IUPAC Name 6-oxoheptanoic acid
SMILES CC(=O)CCCCC(=O)O

Specifications

Melting Point 35°C to 37°C
Boiling Point 138°C to 140°C (1.5 mmHg)
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Linear Formula CH3C(O)(CH2)4CO2H
Quantity 5 g
Solubility Information Solubility in water: soluble
Formula Weight 144.17
Percent Purity 90+%
Grade Technical
Chemical Name or Material 5-Acetylvaleric acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 221-512-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.