Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Allyl-2-hydroxy-3-methoxybenzaldehyde, 98%, Acros Organics

Click to view available options
Quantity:
10g
25g
Packaging:
Glass bottle
22934-51-6
C11H12O3
192.21
MFCD06408022
FDHXEIOBIOVBEN-UHFFFAOYSA-N
5-allyl-2-hydroxy-3-methoxybenzaldehyde,5-allyl-3-methoxysalicylaldehyde,benzaldehyde, 2-hydroxy-3-methoxy-5-2-propenyl,benzaldehyde,2-hydroxy-3-methoxy-5-2-propen-1-yl,2-hydroxy-3-methoxy-5-prop-2-en-1-yl benzaldehyde,formyl eugenol,acmc-209fzq,2-hydroxy-3-methoxy-5-2-propenyl benzaldehyde,3-methoxy-2-oxidanyl-5-prop-2-enyl-benzaldehyde
1713887
2-hydroxy-3-methoxy-5-prop-2-enylbenzaldehyde
COC1=C(C(=CC(=C1)CC=C)C=O)O
22934-51-6
C11H12O3
192.21
MFCD06408022
FDHXEIOBIOVBEN-UHFFFAOYSA-N
5-allyl-2-hydroxy-3-methoxybenzaldehyde,5-allyl-3-methoxysalicylaldehyde,benzaldehyde, 2-hydroxy-3-methoxy-5-2-propenyl,benzaldehyde,2-hydroxy-3-methoxy-5-2-propen-1-yl,2-hydroxy-3-methoxy-5-prop-2-en-1-yl benzaldehyde,formyl eugenol,acmc-209fzq,2-hydroxy-3-methoxy-5-2-propenyl benzaldehyde,3-methoxy-2-oxidanyl-5-prop-2-enyl-benzaldehyde
1713887
2-hydroxy-3-methoxy-5-prop-2-enylbenzaldehyde
COC1=C(C(=CC(=C1)CC=C)C=O)O
Infrared Spectrum Authentic
Color Yellow
Packaging Glass bottle
Melting Point 48.0°C to 52.0°C
Quantity 10g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Solubility Information Solubility in water: insoluble
Formula Weight 192.21
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 5-Allyl-2-hydroxy-3-methoxybenzaldehyde
Show More Show Less