Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-1-(2-hydroxyethyl)pyrazole 99.0+%, TCI America™
SDP

Catalog No. A261925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A261925G 25 g
A26195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A261925G Supplier TCI America Supplier No. A261925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 73616-27-0
Molecular Formula C5H9N3O
Molecular Weight (g/mol) 127.147
MDL Number MFCD08063809
InChI Key IHQRJCVJAUKIEP-UHFFFAOYSA-N
Synonym 5-amino-1-2-hydroxyethyl pyrazole, 2-5-aminopyrazol-1-yl ethanol, 5-amino-1-, a-hydroxyethyl pyrazole, 2-5-amino-1h-pyrazol-1-yl ethanol, 1h-pyrazole-1-ethanol, 5-amino, 2-5-amino-1h-pyrazol-1-yl ethan-1-ol, pubchem15051, ksc496i9l, 2-5-amino-1-pyrazolyl ethanol
PubChem CID 320281
IUPAC Name 2-(5-aminopyrazol-1-yl)ethanol
SMILES C1=C(N(N=C1)CCO)N

Specifications

Melting Point 108°C
Boiling Point 140°C
Quantity 25 g
Formula Weight 127.15
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-1-(2-hydroxyethyl)pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.