Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-1-indanone, 95%, Thermo Scientific™

Catalog No. AAH6642503
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3470-54-0
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00099466
InChI Key HODOSJNSRPXYBH-UHFFFAOYSA-N
Synonym 5-aminoindan-1-one, 5-amino-1-indanone, 5-amino-2,3-dihydro-1h-inden-1-one, 1h-inden-1-one, 5-amino-2,3-dihydro, 5-aminoindanone, 5-amino-1-oxoindane, 5-amino-indan-1-one, pubchem14476, 5-amino-1-indanone #, 5-azanyl-2,3-dihydroinden-1-one
PubChem CID 312890
IUPAC Name 5-amino-2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=C1C=C(C=C2)N

Specifications

Melting Point 184°C to 186°C
Quantity 1 g
UN Number UN2811
Beilstein 1636158
Formula Weight 147.18
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-Amino-1-indanone
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.