Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Amino-1-methylbenzimidazole, 97%

Click to view available options
Quantity:
1g
5g
10394-38-4
C8H9N3
147.181
MFCD03164349
IWBGBYZGEQUDBT-UHFFFAOYSA-N
1-methylbenzoimidazol-5-amine,1-methyl-1h-benzo d imidazol-5-amine,1-methyl-1h-benzimidazol-5-amine,1-methyl-1h-benzoimidazol-5-ylamine,1-methyl-1h-1,3-benzodiazol-5-amine,1-methyl-1,3-benzodiazol-5-amine,1-methyl-1h-benzimidazol-5-ylamine,1-methylbenzimidazole-5-ylamine,1-methyl-5-aminobenzimidazole,5-amino-1-methylbenzimidazole
315499
1-methylbenzimidazol-5-amine
CN1C=NC2=C1C=CC(=C2)N
10394-38-4
C8H9N3
147.181
MFCD03164349
IWBGBYZGEQUDBT-UHFFFAOYSA-N
1-methylbenzoimidazol-5-amine,1-methyl-1h-benzo d imidazol-5-amine,1-methyl-1h-benzimidazol-5-amine,1-methyl-1h-benzoimidazol-5-ylamine,1-methyl-1h-1,3-benzodiazol-5-amine,1-methyl-1,3-benzodiazol-5-amine,1-methyl-1h-benzimidazol-5-ylamine,1-methylbenzimidazole-5-ylamine,1-methyl-5-aminobenzimidazole,5-amino-1-methylbenzimidazole
315499
1-methylbenzimidazol-5-amine
CN1C=NC2=C1C=CC(=C2)N
Quantity 5g
Beilstein 128912
Formula Weight 147.08
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 5-Amino-1-methylbenzimidazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning