Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-1-pentanol, 50 wt.% aqueous solution

Catalog No. p-3212194
Click to view available options
Quantity:
25 mL
100 mL
800 mL
Packaging:
Glass bottle

CAS: 2508-29-4 | C5H13NO | 103.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2508-29-4, 7732-18-5
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.16
MDL Number MFCD00008237
InChI Key LQGKDMHENBFVRC-UHFFFAOYSA-N
Synonym 5-amino-1-pentanol, 5-aminopentanol, 1-pentanol, 5-amino, pentanol, 5-amino, 5-amino-1-pentanol in water, 5-amino pentanol, 5-amino-l-pentanol, 5-aminopentanol-1, 5-hydroxypentylamine, 1-amino-5-pentanol
PubChem CID 75634
IUPAC Name 5-aminopentan-1-ol
SMILES C(CCN)CCO

Specifications

CAS Min % 48.0
CAS Max % 52.0
Density 0.9870g/mL
Flash Point >100°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula OH(CH2)5NH2
Quantity 25 mL
Specific Gravity 0.987
Solubility Information Solubility in water: miscible.
Formula Weight 103.16
Percent Purity 48.0 to 52.0%
Chemical Name or Material 5-Amino-1-pentanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 219-718-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.