Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-1-pentanol 95.0+%, TCI America™
SDP

Catalog No. A0875250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0875250G 250 g
A087525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0875250G Supplier TCI America Supplier No. A0875250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2508-29-4
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.165
MDL Number MFCD00008237
InChI Key LQGKDMHENBFVRC-UHFFFAOYSA-N
Synonym 5-amino-1-pentanol, 5-aminopentanol, 1-pentanol, 5-amino, pentanol, 5-amino, 5-amino-1-pentanol in water, 5-amino pentanol, 5-amino-l-pentanol, 5-aminopentanol-1, 5-hydroxypentylamine, 1-amino-5-pentanol
PubChem CID 75634
IUPAC Name 5-aminopentan-1-ol
SMILES C(CCN)CCO

Specifications

Color White-Yellow
Boiling Point 222°C
Quantity 250 g
UN Number 1759
Formula Weight 103.17
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 5-Amino-1-pentanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.