Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-1H-benzimidazole-2-thiol, 97%, Thermo Scientific™

Catalog No. AC34641DA
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2818-66-8
Molecular Formula C7H7N3S
Molecular Weight (g/mol) 165.21
MDL Number MFCD00022671
InChI Key BXDMTLVCACMNJO-UHFFFAOYSA-N
Synonym 5-amino-2-mercaptobenzimidazole, 5-amino-2-benzimidazolethiol, 5-amino-1h-benzo d imidazole-2-thiol, 5-amino-1h-benzimidazole-2-thiol, 5-aminobenzimidazole-2-thiol, 2h-benzimidazole-2-thione, 5-amino-1,3-dihydro, 2-benzimidazolethiol, 5-amino, 5-amino-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-amino benzimidazole, 5-amino-3h-1,3-benzodiazole-2-thiol
PubChem CID 2734002
SMILES NC1=CC=C2NC(=S)NC2=C1

Specifications

Assay Percent Range 97%
Quantity 1 g
IUPAC Name 5-amino-1,3-dihydrobenzimidazole-2-thione
Formula Weight 165.22
Chemical Name or Material 5-Amino-1H-benzimidazole-2-thiol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.