Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2,4-dichlorophenol, 97%, Thermo Scientific™

Catalog No. AAH6675303
Click to view available options
Quantity:
250 mg
1 g
5 g

Catalog No. AAH6675303

Supplier: Thermo Scientific Chemicals H6675303

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39489-79-7
Molecular Formula C6H5Cl2NO
Molecular Weight (g/mol) 178.01
MDL Number MFCD00186187
InChI Key QPHMVRPABQUYGN-UHFFFAOYSA-N
PubChem CID 101471
IUPAC Name 5-amino-2,4-dichlorophenol
SMILES NC1=CC(O)=C(Cl)C=C1Cl

Specifications

Melting Point 133°C to 137°C
Quantity 1 g
UN Number UN2811
Beilstein 2717129
Synonym 2,4-dichloro-5-aminophenol, 5-amino-2,4-dichlorobenzenol, 2,4-dichloro-5-hydroxyaniline, phenol, 5-amino-2,4-dichloro, aminodichlorobenzenol, acmc-20aidn, 3-amino-4,6-dichlorophenol, 5-amino-2,4-dichloro-phenol, phenol,5-amino-2,4-dichloro
Formula Weight 178.01
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Amino-2,4-dichlorophenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.