Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-(4-morpholinyl)pyridine, 97%

Catalog No. AAH5069903
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 52023-68-4 | C9H13N3O | 179.223 g/mol

5-Amino-2-(4-morpholinyl)pyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Amino-2-(4-morpholinyl)pyridine is used as pharmaceutical intermediate.

Notes
Store at room temperature. Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52023-68-4
Molecular Formula C9H13N3O
Molecular Weight (g/mol) 179.223
MDL Number MFCD00119562
InChI Key VVTSPTCBHTWXMD-UHFFFAOYSA-N
Synonym 6-morpholinopyridin-3-amine, 3-amino-6-morpholinopyridine, 6-morpholino-3-pyridinamine, 6-morpholin-4-yl pyridin-3-amine, 6-morpholino pyridin-3-amine, 6-morpholinopyridine-3-amine, 3-pyridinamine, 6-4-morpholinyl, 6-morpholin-4-yl-3-pyridylamine, 5-amino-2-morpholin-4-ylpyridine, 5-amino-2-4-morpholinyl pyridine
PubChem CID 104059
IUPAC Name 6-morpholin-4-ylpyridin-3-amine
SMILES C1COCCN1C2=NC=C(C=C2)N

Specifications

Melting Point 127°C to 130°C
Quantity 1 g
Formula Weight 179.22
Percent Purity 97%
Chemical Name or Material 5-Amino-2-(4-morpholinyl)pyridine
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 257-613-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.