Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Amino-2-bromo-6-methylpyridine, 98%

Click to view available options
Quantity:
1g
5g
25g
126325-47-1
C6H7BrN2
187.04
MFCD03095086
UBTQJSZELCWGCN-UHFFFAOYSA-N
5-amino-2-bromo-6-picoline,3-amino-6-bromo-2-methylpyridine,2-bromo-5-amino-6-methylpyridine,3-amino-6-bromo-2-picoline,5-amino-2-bromo-6-methylpyridine,2-bromo-5-amino-6-picoline,6-bromo-2-methyl-3-pyridylamine,6-bromo-2-methyl-pyridin-3-ylamine,2-methyl-3-amino-6-bromopyridine,pubchem1117
14418045
6-bromo-2-methylpyridin-3-amine
CC1=C(C=CC(=N1)Br)N
126325-47-1
C6H7BrN2
187.04
MFCD03095086
UBTQJSZELCWGCN-UHFFFAOYSA-N
5-amino-2-bromo-6-picoline,3-amino-6-bromo-2-methylpyridine,2-bromo-5-amino-6-methylpyridine,3-amino-6-bromo-2-picoline,5-amino-2-bromo-6-methylpyridine,2-bromo-5-amino-6-picoline,6-bromo-2-methyl-3-pyridylamine,6-bromo-2-methyl-pyridin-3-ylamine,2-methyl-3-amino-6-bromopyridine,pubchem1117
14418045
6-bromo-2-methylpyridin-3-amine
CC1=C(C=CC(=N1)Br)N
Melting Point 110°C to 111°C
Quantity 1g
Color White
Beilstein 6190039
Formula Weight 187.04
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 5-Amino-2-bromo-6-methylpyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning