Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-chloro-3-methylpyridine 97.0+%, TCI America™
SDP

Catalog No. A25645G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 38186-82-2
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD03095087
InChI Key VSBISZPNLZFTPG-UHFFFAOYSA-N
Synonym 5-amino-2-chloro-3-picoline, 2-chloro-3-methyl-5-aminopyridine, 5-amino-2-chloro-3-methylpyridine, 3-pyridinamine, 6-chloro-5-methyl, 6-chloro-5-methyl-3-pyridylamine, 6-chloro-5-methyl-pyridin-3-ylamine, 6-chloro-5-methylpyridine-3-amine, pubchem1152, 5-methyl-6-chloropyridine-3-amine, 6-chloro-5-methyl-3-pyridinamine
PubChem CID 12295607
IUPAC Name 6-chloro-5-methylpyridin-3-amine
SMILES CC1=CC(N)=CN=C1Cl

Specifications

Melting Point 93°C
Quantity 5 g
Formula Weight 142.59
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-chloro-3-methylpyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.