Learn More
5-Amino-2-chlorobenzoic acid, 97%, Thermo Scientific™
Specifications
Specifications
| Melting Point | 184.0°C to 188.0°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (HPLC) |
| Quantity | 100 g |
| Formula Weight | 171.58 |
| Percent Purity | 97% |
| Chemical Name or Material | 5-Amino-2-chlorobenzoic acid |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF |
| EINECSNumber | 201-916-5 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.