Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Amino-2-methylbenzothiazole, 99%

Click to view available options
Quantity:
2g
10g
50g
13382-43-9
C8H8N2S
164.226
MFCD00226291
GPWQHYMVUZYWIK-UHFFFAOYSA-N
2-methylbenzo d thiazol-5-amine,5-amino-2-methylbenzothiazole,5-benzothiazolamine, 2-methyl,2-methyl-5-benzothiazolamine,2-methyl-5-aminobenzothiazole,2-methylbenzothiazole-5-ylamine,2-methyl-1,3-benzothiazol-5-ylamine,acmc-209v5w,cambridge id 5107842,2-methylbenzothiazol-5-amine
36229
2-methyl-1,3-benzothiazol-5-amine
CC1=NC2=C(S1)C=CC(=C2)N
13382-43-9
C8H8N2S
164.226
MFCD00226291
GPWQHYMVUZYWIK-UHFFFAOYSA-N
2-methylbenzo d thiazol-5-amine,5-amino-2-methylbenzothiazole,5-benzothiazolamine, 2-methyl,2-methyl-5-benzothiazolamine,2-methyl-5-aminobenzothiazole,2-methylbenzothiazole-5-ylamine,2-methyl-1,3-benzothiazol-5-ylamine,acmc-209v5w,cambridge id 5107842,2-methylbenzothiazol-5-amine
36229
2-methyl-1,3-benzothiazol-5-amine
CC1=NC2=C(S1)C=CC(=C2)N
Applications
5-Amino-2-methylbenzothiazole is used as pharmaceutical intermediates.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Quantity 2g
Formula Weight 164.22
Percent Purity 99%
Chemical Name or Material 5-Amino-2-methylbenzothiazole

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA