Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Aminoindan 98.0+%, TCI America™
SDP

Catalog No. A099125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A099125G 25 g
A099110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A099125G Supplier TCI America Supplier No. A099125G

Chemical Identifiers

CAS 24425-40-9
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.194
MDL Number MFCD00003803
InChI Key LEWZOBYWGWKNCK-UHFFFAOYSA-N
Synonym 5-aminoindan, indan-5-amine, indan-5-ylamine, 5-indanamine, 5-aminohydrindene, 5-aminoindane, indane-5-ylamine, 1h-inden-5-amine, 2,3-dihydro, 2,3-dihydro-1h-inden-5-ylamine, 6-aminoindane
PubChem CID 90496
IUPAC Name 2,3-dihydro-1H-inden-5-amine
SMILES C1CC2=C(C1)C=C(C=C2)N

Specifications

Melting Point 37°C
Boiling Point 147°C
Quantity 25 g
Formula Weight 133.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminoindan
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.