Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Aminophthalide, 97%

Click to view available options
Quantity:
1g
5g
65399-05-5
C8H7NO2
149.149
MFCD00778315
ISMUWQMUWFPFBZ-UHFFFAOYSA-N
5-aminophthalide,5-aminoisobenzofuran-1 3h-one,5-amino-3h-isobenzofuran-1-one,5-amino-2-benzofuran-1 3h-one,1 3h-isobenzofuranone, 5-amino,5-amino-1 3h-isobenzofuranone,5-amino-1,3-dihydro-2-benzofuran-1-one,5-amino-3-hydroisobenzofuran-1-one,5-aminophthliade,5-amino-phthalide
720669
5-amino-3H-2-benzofuran-1-one
C1C2=C(C=CC(=C2)N)C(=O)O1
65399-05-5
C8H7NO2
149.149
MFCD00778315
ISMUWQMUWFPFBZ-UHFFFAOYSA-N
5-aminophthalide,5-aminoisobenzofuran-1 3h-one,5-amino-3h-isobenzofuran-1-one,5-amino-2-benzofuran-1 3h-one,1 3h-isobenzofuranone, 5-amino,5-amino-1 3h-isobenzofuranone,5-amino-1,3-dihydro-2-benzofuran-1-one,5-amino-3-hydroisobenzofuran-1-one,5-aminophthliade,5-amino-phthalide
720669
5-amino-3H-2-benzofuran-1-one
C1C2=C(C=CC(=C2)N)C(=O)O1
Quantity 1g
Beilstein 121983
Formula Weight 149.05
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 5-Aminophthalide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning