Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Aminopyrazine-2-carboxylic acid, 95%

Catalog No. p-7056955
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 40155-43-9 | C5H5N3O2 | 139.114 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40155-43-9
Molecular Formula C5H5N3O2
Molecular Weight (g/mol) 139.114
MDL Number MFCD07371664
InChI Key FMEFOOOBIHQRMK-UHFFFAOYSA-N
Synonym 5-amino-2-pyrazinecarboxylic acid, 2-amino-5-pyrazinecarboxylic acid, pyrazinecarboxylic acid, 5-amino, 5-amino-pyrazine-2-carboxylic acid, 5-aminopyrazine-2-carboxylicacid, 5-amino-pyrazine-2-carboxylicacid, pubchem7866, 5-amino-pyrazinecarboxylic acid, 2-amino-5-pirazinecarboxylic acid
PubChem CID 21897334
IUPAC Name 5-aminopyrazine-2-carboxylic acid
SMILES C1=C(N=CC(=N1)N)C(=O)O

Specifications

Quantity 250 mg
Beilstein 120673
Formula Weight 139.11
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-Aminopyrazine-2-carboxylic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.