Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Aminoquinoline 99.0+%, TCI America™
SDP

Catalog No. A095910G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
10 g
25 g

Chemical Identifiers

CAS 611-34-7
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00006797
InChI Key XMIAFAKRAAMSGX-UHFFFAOYSA-N
Synonym 5-aminoquinoline, 5-quinolinamine, quinoline, 5-amino, 5-quinolylamine, quinolin-5-ylamine, unii-yyj34z13sh, 5-amino-quinoline, ccris 1680, yyj34z13sh
PubChem CID 11911
IUPAC Name quinolin-5-amine
SMILES NC1=C2C=CC=NC2=CC=C1

Specifications

Melting Point 109°C
Boiling Point 310°C
Quantity 10 g
Formula Weight 144.18
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminoquinoline
RTECSNumber VA9625000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.