Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Aminovaleric acid, 97%

Catalog No. ac153910050
Click to view available options
Quantity:
5 g
25 g

CAS: 660-88-8 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 660-88-8
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00008232
InChI Key JJMDCOVWQOJGCB-UHFFFAOYSA-N
Synonym 5-aminovaleric acid, 5-amino-pentanoic acid, homopiperidinic acid, 5-aminopentanoate, valeric acid, 5-amino, delta-amino-n-valeric acid, pentanoic acid, 5-amino, delta-aminovaleric acid, 5-amino-n-valeric acid, 5-aminovalerate
PubChem CID 138
ChEBI CHEBI:15887
IUPAC Name 5-aminopentanoic acid
SMILES C(CCN)CC(=O)O

Specifications

Melting Point 158.0°C to 161.0°C
CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula H2N(CH2)4CO2H
Beilstein 04, 418
Solubility Information solution
Quantity 5 g
Formula Weight 117.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminovaleric acid, 97%
Show More Show Less
EINECSNumber 211-544-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.