Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Azacytosine 98.0+%, TCI America™
SDP

Catalog No. A109125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A109125G 25 g
A10915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A109125G Supplier TCI America Supplier No. A109125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 931-86-2
Molecular Formula C3H4N4O
Molecular Weight (g/mol) 112.09
MDL Number MFCD00006033,MFCD00149402,MFCD00051007
InChI Key MFEFTTYGMZOIKO-UHFFFAOYSA-N
Synonym 5-azacytosine, 1,3,5-triazin-2 1h-one, 4-amino, 4-amino-1,3,5-triazin-2 1h-one, 4-amino-1,3,5-triazin-2-ol, 4-amino-1,3,5-triazin-2-one, s-triazin-2 1h-one, 4-amino, s-triazin-2-ol, 4-amino, 2-amino-4-hydroxy-s-triazine, 6-amino-1,3,5-triazin-2 1h-one, 4-amino-s-triazin-2 1h-one
PubChem CID 19956
ChEBI CHEBI:72474
IUPAC Name 6-amino-1,2-dihydro-1,3,5-triazin-2-one
SMILES NC1=NC=NC(=O)N1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 112.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Azacytosine
RTECSNumber XZ2854300
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.