Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Azaindole, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
271-34-1
C7H6N2
118.14
SRSKXJVMVSSSHB-UHFFFAOYSA-N
5-azaindole,1h-pyrrolo 3,2-c pyridine,1,5-diazaindene,3,7-dideazapurine,5-azindole,5h-pyrrolo 3,2-c pyridine,pyrrolo 3,2-c pyridine,5-aza indole,5-aza-indole
9220
1H-pyrrolo[3,2-c]pyridine
C1=CNC2=C1C=NC=C2
271-34-1
C7H6N2
118.14
SRSKXJVMVSSSHB-UHFFFAOYSA-N
5-azaindole,1h-pyrrolo 3,2-c pyridine,1,5-diazaindene,3,7-dideazapurine,5-azindole,5h-pyrrolo 3,2-c pyridine,pyrrolo 3,2-c pyridine,5-aza indole,5-aza-indole
9220
1H-pyrrolo[3,2-c]pyridine
C1=CNC2=C1C=NC=C2
Melting Point 93.0°C to 94.0°C
Quantity 5g
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 118.14
Percent Purity 98%
Chemical Name or Material 5-Azaindole

GHS H Statement
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning