Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Benzyl-1H-tetrazole, 99%, Thermo Scientific Chemicals

Catalog No. p-7033544
Click to view available options
Quantity:
1 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18489-25-3
Molecular Formula C8H8N4
Molecular Weight (g/mol) 160.18
MDL Number MFCD00068730
InChI Key HHDRWGJJZGJSGZ-UHFFFAOYSA-N
Synonym 5-benzyl-1h-tetrazole, 5-benzyltetrazole, 1h-tetrazole, 5-phenylmethyl, 5-benzyl-2h-1,2,3,4-tetraazole, 5-benzyl-1h-1,2,3,4-tetrazole, 5-benzyl-2h-1,2,3,4-tetrazole, 5-phenylmethyltetrazole, maybridge1_003729, acmc-209em5, 5-phenylmethyl-1h-tetrazole
PubChem CID 223451
IUPAC Name 5-benzyl-2H-tetrazole
SMILES C(C1=NNN=N1)C1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 160.18
Percent Purity 99%
Chemical Name or Material 5-Benzyl-1H-tetrazole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.