Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Benzyloxy-2-bromo-4-methoxybenzoic Acid 97.0+%, TCI America™
SDP

Catalog No. B33321G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33321G 1 g
B33325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33321G Supplier TCI America Supplier No. B33321G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24958-42-7
Molecular Formula C15H13BrO4
Molecular Weight (g/mol) 337.17
MDL Number MFCD03695499
InChI Key FBTDEMXQQIWHKX-UHFFFAOYSA-N
Synonym 5-Benzyloxy-2-bromo-p-anisic Acid
PubChem CID 11056839
IUPAC Name 5-(benzyloxy)-2-bromo-4-methoxybenzoic acid
SMILES COC1=C(OCC2=CC=CC=C2)C=C(C(O)=O)C(Br)=C1

Specifications

Melting Point 195°C
Quantity 1 g
Formula Weight 337.17
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Benzyloxy-2-bromo-4-methoxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.