Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 5-BrdU
GREENER_CHOICESDP

Catalog No. 501550R Shop All Novus Biologicals Products
Change view
Click to view available options
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
501550R 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 501550R Supplier Tocris Bioscience™ Supplier No. 5015/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Synthetic thymidine analog; used for cell proliferation assays

5-BrdU is a synthetic thymidine analog; incorporated into DNA during replication. Used in assays for cell proliferation. Replaces Oct-4 in transcription factor-mediated reprogramming of somatic cells and can be used in small molecule cocktail to generate ciPSCs.

Specifications

Quantity 50 mg
Product Type 5-BrdU
Purity >99%
Molecular Weight (g/mol) 307.1
Inhibitors 5-BrdU

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.