Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-1-benzothiophene, 97%, Thermo Scientific™

Catalog No. CC24810DA
Click to view available options
:
Amber Glass Bottle; 1g

Specifications

CAS 4923-87-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H5BrS
Quantity 1g
MDL Number MFCD03069318
Synonym 5-bromobenzo b thiophene, 5-bromobenzothiophene, 5-bromo-benzo b thiophene, 5-bromothianaphthene, benzo b thiophene, 5-bromo, zlchem 181, pubchem7752, 5-bromo-benzothiophene, pubchem15353, acmc-1anox
InChI Key RDSIMGKJEYNNLF-UHFFFAOYSA-N
SMILES BrC1=CC=C2SC=CC2=C1
IUPAC Name 5-bromo-1-benzothiophene
Molecular Weight (g/mol) 213.09
PubChem CID 2776578
Formula Weight 213.1
Percent Purity 97%
Chemical Name or Material 5-Bromo-1-benzothiophene
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.