Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-1-methylisatin 98.0+%, TCI America™
SDP

Catalog No. B47791G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47791G 1 g
B47795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47791G Supplier TCI America Supplier No. B47791G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2058-72-2
Molecular Formula C9H6BrNO2
Molecular Weight (g/mol) 240.06
MDL Number MFCD00456313
InChI Key GEEDYJPPYNIZLX-UHFFFAOYSA-N
Synonym 5-Bromo-1-methylindole-2,3-dione, 5-Bromo-1-methyl-2,3-indolinedione
PubChem CID 16357
IUPAC Name 5-bromo-1-methyl-2,3-dihydro-1H-indole-2,3-dione
SMILES CN1C(=O)C(=O)C2=CC(Br)=CC=C12

Specifications

Melting Point 171°C
Color Red-Yellow
Quantity 1 g
Formula Weight 240.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-1-methylisatin
RTECSNumber NL7878000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.