Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-1H-indazole-3-carboxylic acid, 95%, Thermo Scientific™

Catalog No. MO07350DE
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 1077-94-7
Molecular Formula C8H5BrN2O2
Molecular Weight (g/mol) 241.04
MDL Number MFCD05663979
InChI Key AMJVXOOGGBPVCZ-UHFFFAOYSA-N
PubChem CID 7157358
IUPAC Name 5-bromo-1H-indazole-3-carboxylic acid
SMILES OC(=O)C1=NNC2=CC=C(Br)C=C12

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 5-bromo-1h-indazole-3-carboxylicacid, 5-bromoindazole-3-carboxylic acid, 5-bromo-3-indazolecarboxylic acid, 1h-indazole-3-carboxylic acid, 5-bromo, 1h-indazole-3-carboxylic acid,5-bromo, pubchem7817, acmc-2098xf, ksc173q5b, 5-bromo-3-carboxy-1h-indazole, 1h-indazole-3-carboxylicacid, 5-bromo
Formula Weight 241.04
Percent Purity 95%
Chemical Name or Material 5-Bromo-1H-indazole-3-carboxylic acid
Hazard Category Warning
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.