Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5″-Bromo-2,2':5',2″-terthiophene-5-carboxaldehyde 94.0+%, TCI America™
SDP

Catalog No. B2058100MG
Click to view available options
Quantity:
100 mg
1 g
5 g

Chemical Identifiers

CAS 161726-69-8
Molecular Formula C13H7BrOS3
Molecular Weight (g/mol) 355.28
MDL Number MFCD02093442
InChI Key DSYKMLTVEGWLFR-UHFFFAOYSA-N
PubChem CID 14942412
IUPAC Name 5-{5'-bromo-[2,2'-bithiophen]-5-yl}thiophene-2-carbaldehyde
SMILES BrC1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C=O

Specifications

Melting Point 164°C
Quantity 100 mg
Synonym 5-Bromo-5′C′C-formyl-2,2′C:5′C,2′C′C-terthiophene
Formula Weight 355.28
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5′′-Bromo-2,2′:5′,2′′-terthiophene-5-carboxaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.