Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5'-Bromo-[2,2'-bithiophene]-5-carbonitrile 98.0+%, TCI America™
SDP

Catalog No. B54571G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54571G 1 g
B5457200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B54571G Supplier TCI America Supplier No. B54571G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 176787-96-5
Molecular Formula C9H4BrNS2
Molecular Weight (g/mol) 270.16
MDL Number MFCD16339405
InChI Key OHVCHDZJRUEOKH-UHFFFAOYSA-N
PubChem CID 15458991
IUPAC Name 5'-bromo-[2,2'-bithiophene]-5-carbonitrile
SMILES BrC1=CC=C(S1)C1=CC=C(S1)C#N

Specifications

Melting Point 163°C
Quantity 1 g
Formula Weight 270.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5′-Bromo-[2,2′-bithiophene]-5-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.