Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5'-Bromo-[2,2'-bithiophene]-5-carbonitrile 98.0+%, TCI America™
SDP

Catalog No. p-7124083
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 176787-96-5
Molecular Formula C9H4BrNS2
Molecular Weight (g/mol) 270.16
MDL Number MFCD16339405
InChI Key OHVCHDZJRUEOKH-UHFFFAOYSA-N
PubChem CID 15458991
IUPAC Name 5'-bromo-[2,2'-bithiophene]-5-carbonitrile
SMILES BrC1=CC=C(S1)C1=CC=C(S1)C#N

Specifications

Melting Point 163°C
Quantity 200 mg
Formula Weight 270.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5′-Bromo-[2,2′-bithiophene]-5-carbonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.