Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2,4-dimethoxypyrimidine 98.0+%, TCI America™
SDP

Catalog No. B192025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B192025G 25 g
B19201G 1 g
B19205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B192025G Supplier TCI America Supplier No. B192025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56686-16-9
Molecular Formula C6H7BrN2O2
Molecular Weight (g/mol) 219.038
MDL Number MFCD00038016
InChI Key QEZIMQMMEGPYTR-UHFFFAOYSA-N
Synonym 2,4-dimethoxy-5-bromopyrimidine, pyrimidine, 5-bromo-2,4-dimethoxy, 5-bromo-2,4-dimethoxy-1,3-diazine, 5-bromo-2,4-bis methyloxy pyrimidine, 5-bromo-2,4-dimethoxy-pyrimidine, pubchem5281, acmc-209lu1, ksc495s0l, 5-bromo-2,4-dimethoxypyrimidine, 5-bromo-2,4-dimethoxypyrimidine;
PubChem CID 255719
IUPAC Name 5-bromo-2,4-dimethoxypyrimidine
SMILES COC1=NC(=NC=C1Br)OC

Specifications

Melting Point 63°C
Color White-Yellow
Boiling Point 125°C
Quantity 25 g
Formula Weight 219.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2,4-dimethoxypyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.