Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-chlorobenzonitrile 98.0+%, TCI America™
SDP

Catalog No. B311625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B311625G 25 g
B31165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B311625G Supplier TCI America Supplier No. B311625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57381-44-9
Molecular Formula C7H3BrClN
Molecular Weight (g/mol) 216.462
MDL Number MFCD00672948
InChI Key NKDMQBUJOXQVEA-UHFFFAOYSA-N
PubChem CID 21525431
IUPAC Name 5-bromo-2-chlorobenzonitrile
SMILES C1=CC(=C(C=C1Br)C#N)Cl

Specifications

Melting Point 133°C
Color White-Yellow
Quantity 25 g
UN Number 3439
Formula Weight 216.46
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-chlorobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.