Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-chloropyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
53939-30-3
C5H3BrClN
192.44
MFCD01318951
PEAOEIWYQVXZMB-UHFFFAOYSA-N
2-chloro-5-bromopyridine,5-bromo-2-chloro-pyridine,pyridine, 5-bromo-2-chloro,2-chloro-5-bromo-pyridine,2-chlorine-5-bromine pyridine,pubchem1168,acmc-1afhf,3-bromo-6-chloropyridine,2-chloro-5-bromo pyridine,3-bromo-6-chloro-pyridine
2734414
5-bromo-2-chloropyridine
C1=CC(=NC=C1Br)Cl
53939-30-3
C5H3BrClN
192.44
MFCD01318951
PEAOEIWYQVXZMB-UHFFFAOYSA-N
2-chloro-5-bromopyridine,5-bromo-2-chloro-pyridine,pyridine, 5-bromo-2-chloro,2-chloro-5-bromo-pyridine,2-chlorine-5-bromine pyridine,pubchem1168,acmc-1afhf,3-bromo-6-chloropyridine,2-chloro-5-bromo pyridine,3-bromo-6-chloro-pyridine
2734414
5-bromo-2-chloropyridine
C1=CC(=NC=C1Br)Cl
Melting Point 67°C to 71°C
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Color White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 192.44
Physical Form Crystalline Powder or Crystals
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-chloropyridine
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning