Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-chloropyrimidine, 96%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass bottle
32779-36-5
C4H2BrClN2
193.43
MFCD00483232
XPGIBDJXEVAVTO-UHFFFAOYSA-N
2-chloro-5-bromopyrimidine,5-bromo-2-chloro-pyrimidine,5-bromo-2-chloropyrimidne,pyrimidine, 5-bromo-2-chloro,5-bromo-2-chloro-1,3-diazine,pubchem5159,zlchem 1210,acmc-209hvm,5-bromo chloropyrimidine,5-bromo-2-chloroprimidine
606665
5-bromo-2-chloropyrimidine
C1=C(C=NC(=N1)Cl)Br
32779-36-5
C4H2BrClN2
193.43
MFCD00483232
XPGIBDJXEVAVTO-UHFFFAOYSA-N
2-chloro-5-bromopyrimidine,5-bromo-2-chloro-pyrimidine,5-bromo-2-chloropyrimidne,pyrimidine, 5-bromo-2-chloro,5-bromo-2-chloro-1,3-diazine,pubchem5159,zlchem 1210,acmc-209hvm,5-bromo chloropyrimidine,5-bromo-2-chloroprimidine
606665
5-bromo-2-chloropyrimidine
C1=C(C=NC(=N1)Cl)Br
Quantity 250mg
Infrared Spectrum Authentic
Melting Point 78.0°C to 80.0°C
Packaging Glass bottle
Color White to Beige
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (HPLC)
Solubility Information Solubility in water: insoluble in water
Formula Weight 193.43
Physical Form Crystalline Powder
Percent Purity 96%
Chemical Name or Material 5-Bromo-2-chloropyrimidine
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning