Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-Bromo-2-chlorotoluene, 98%, Thermo Scientific™
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Specifications
Specifications
| Color | Colorless to Orange-Yellow |
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Density | 1.5490g/mL |
| Boiling Point | 90.0°C to 93.0°C (12.0 mmHg) |
| Quantity | 5 g |
| Flash Point | >110°C |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction