Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-cyano-3-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. B558425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B558425G 25 g
B55845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B558425G Supplier TCI America Supplier No. B558425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 573675-25-9
Molecular Formula C6H2BrN3O2
Molecular Weight (g/mol) 228.005
MDL Number MFCD06657551
InChI Key OBVYONPVHIOJJZ-UHFFFAOYSA-N
Synonym 5-Bromo-3-nitropyridine-2-carbonitrile, 5-Bromo-3-nitropicolinonitrile
PubChem CID 2769698
IUPAC Name 5-bromo-3-nitropyridine-2-carbonitrile
SMILES C1=C(C(=NC=C1Br)C#N)[N+](=O)[O-]

Specifications

Melting Point 107°C
Color Yellow
Quantity 25 g
UN Number 3439
Formula Weight 228.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-cyano-3-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.