Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-ethynylpyridine 98.0+%, TCI America™
SDP

Catalog No. B51791G
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 111770-86-6
Molecular Formula C7H4BrN
Molecular Weight (g/mol) 182.02
MDL Number MFCD03095276
InChI Key VGLAJERHBBEPQP-UHFFFAOYSA-N
PubChem CID 13979734
IUPAC Name 5-bromo-2-ethynylpyridine
SMILES BrC1=CN=C(C=C1)C#C

Specifications

Melting Point 85°C
Quantity 1 g
Formula Weight 182.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-ethynylpyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.